Learn More
Sino Biological Recombinant Mouse Fas Ligand/TNFSF6 Protein (His Tag)
Supplier:  Sino Biological 50854M07Y50
Encompass_Preferred
A DNA sequence encoding the mouse Faslg (AAB02915.1) (Pro132-Leu279) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-2586