Learn More
Alkali Scientific Conical Centrifuge Tubes, 500mL, PP, Plug Seal Cap, Sterile, 6/Bag, 36/cs
SDP
Supplier:  Alkali Scientific C8250
Encompass
The 500mL conical centrifuge tube can process a large number of samples at one time and are suitable for separation and harvest of large cells, supernatants, bacteria, yeast and tissue samples, therefore they are widely used in the field of life sciences and clinical experiments and can meet the requirements of biological analysis. The unique sealing structure design of the cover provides a two-way security to guarantee seal ability. They can tolerate temperatures of -80℃ to 120℃. Sterlie, DNase, RNase, and pyrogen-free.
Catalog No. 50-233-5824