Learn More
Bellco Glass, Inc. 50ml Centrifuge Tube, PS Rack, standard, sterile 25/pk, 500/cs
SDP
Supplier:  Bellco Glass, Inc. 451400277
Encompass_Preferred
50ml Centrifuge Tube, PS Rack, standard, sterile 25/pk, 500/cs. centrifuge tubes feature black printed graduations and a large white marking field.Premium• Meet the requirements of analytical level• High strength non-toxic materials• Raw material comply with USP VI requirementsStandard & Self-standing• Made from virgin polypropylene to meet the general requirements of the laboratory• Made from virgin polypropylene, skirted and self-standing • Printed Graduations (with marking area)• Polypropylene tube and high density polyethylene cap• High centrifugal strength - up to 10,000 x g• Flat markable cap with leakproof seal• One-hand operation design for the caps provides easy handling• Sterilized by E-Beam• DNase, RNase and pyrogen free
Catalog No. 50-233-0265