Learn More
CELLTREAT Scientific Products 50mL Centrifuge Tube, Assorted Color Cap - Bag, Sterile, 500/CS
SDP
Supplier:  CELLTREAT Scientific Products 230421A
Encompass_Preferred
CELLTREAT Scientific Products 50mL Centrifuge Tube, Assorted Color Caps. Graduations are printed in bold, easy-to-read black ink and are accurate to +/-2%. Large white marking area provides excellent contrast for identification. Easy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof seal. Polypropylene tubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantification. Convenient resealable bag is a practical alternative to folding and taping opened bags. Assorted color cap tubes allow easily color coding of samples for quick identification, customization, and increased productivity. Each case of assorted color tubes comes with a free plastic rack, and 4 bags of each individual color (red, yellow, green, blue, purple). Max RCF: 16,000g. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 25/resealable bag, 500/case.
Catalog No. 50-153-5101