Learn More
CELLTREAT Scientific Products 50mL Centrifuge Tube, Snap-Pop Lid - Bag, Sterile, 500/CS
Supplier: CELLTREAT Scientific Products 229498
CELLTREAT Scientific Products 50mL Snap-Pop Lid Centrifuge Tube. Single piece lid and tube with conical bottom. Tab on lid enables convenient one-handed opening. Lid audibly snaps closed, and opens with a 'pop!' so you know when it is sealed. Ideal for sample storage, collection, and experimental procedures. Tube features molded graduation marks at 2.5mL increments, numbered every 5mL. 50mL maximum volume. Temperature range of -80¿C to 120¿C. Max RCF: 9,000g. Tube and lid manufactured from virgin polypropylene. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 25/re-sealable bag, 500/case.