Learn More
Expression Systems Sf9 cells, Suspension in ESF 921, 1M cells/mL (50mL/125mL flask)
SDP
Supplier:  Expression Systems 94001S
Encompass_Preferred
Expression Systems offers insect cell lines adapted to serum-free suspension culture in either ESF 921 or ESF AF. The availability of pre-adapted cells saves significant time and expense associated with the adaptation of cultures.
The Sf9 insect cell line is a clonal isolate derived from the parental Spodoptera frugiperda cell line IPLB-Sf-21-AE. Sf9 cells are adapted for suspension culture in ESF 921 or ESF AF and are available as a frozen vial or suspension culture. Sf9 cells are commonly used to produce recombinant baculoviral stocks and to produce recombinant proteins.
• Recombinant protein expression from a variety of expression platforms
• Good growth in adherent or suspension culture
• Small, regular size that generates even monolayers and plaques
Quality and performance testing
To download Instructions for Use please visit: www.expressionsystems.com
Catalog No. NC0612106
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.