Learn More
Bellco Glass, Inc. 50mL Reagent Reservoir
SDP
Supplier:  Bellco Glass, Inc. 229290000
Encompass_Preferred
DescriptionReagent Reservoirs are designed to assist in repetitive liquid transferWhite color enhances solution visibilityAngled design promotes funneling of liquid to maximize retrieval Pack Description1/BagCase Quantity80SterileYesNon-pyrogenicYesPrimary ColorWhitePrimary MaterialPolystyreneMaximum Volume50mLGraduatedNoRNase/DNase FreeYes
Catalog No. 50-232-9342