Learn More
Chemglass Life Sciences Ultra Tissue Grinder, 50mL, Conical
SDP
Supplier:  Chemglass Life Sciences CLS5150
Encompass_Preferred
Ultra Tissue Grinder, 50mL, Conical. The pestle tips of the new Ultra Tissue Grinders have been vitrified using a proprietary silica casting process. The result is a glass-like abrasive surface which provides a more aggressive grinding action to enable efficient processing of difficult and fibrous samples. Ultra Tissue Grinders incorporate the same patented closed system as the standard grinders. Available in 15mL and 50mL sizes, they are supplied gamma sterilized in an easy-to-open package with an extra cap for storage and transport plus a patient identification label for traceability.
Catalog No. 50-121-4698