Learn More
Across International 50mm Diameter ID 250°C Heated Die w/ Digital Controller
SDP
Supplier:  Across International SDS50.H
Encompass_Preferred
A set of 50mm diameter (ID) hardened steel dry pressing die set for pellet presses, ready for you to make 50mm diameter cylinder-shape pellets. Heating system with programmable digital temperature controller allows heating up to 250C.
Catalog No. 50-225-1322