Learn More
Alkali Scientific Alkali Scientific™ 50uL Beckman™ Biomek FX/NX, Multimek and i Series Compatible Robotic Automation Clear Tips (P50), 96-Well Head/Tip Format, Low Retention, Sterile, Non-Filtered, 10 Racks of 96, 960/Pack
SDP
Supplier:  Alkali Scientific FXS50
Encompass
Crafted from high-quality polypropylene, Alkali Scientific™ provides a full range of tips designed for Beckman Coulter automation systems. These clear tips are produced to exacting standards with rigorous process controls and automated assembly to ensure reliable performance and consistent quality. They are certified free of RNase, DNase, and pyrogens. Designed for compatibility with Beckman Coulter Biomek FX, Biomek NX, Biomek 3000, Multimek AP-96, and i-Series systems, these filtered tips help reduce the risk of sample cross-contamination from aerosols. Like all our tips, the P50s are made from 100% polypropylene, ensuring they are straight, leak-proof, and free of DNA, DNase/RNase, PCR inhibitors, pyrogens/endotoxins, and trace metals. These tips are produced in compliance with ISO 9001/13485, ISO 14001, and CE certifications.
Catalog No. 50-302-7376