Learn More
Trajan Scientific 50 µL removable needle syringe with 5 cm 0.5 mm OD bevel tipped needle
SDP
Supplier:  Trajan Scientific 004050
Encompass_Preferred
50 µL removable needle syringe with 5 cm 0.5 mm OD bevel tipped needle
Catalog No. NC3449135