Learn More
AUTOMATION TRAINER 50 L, Clear, Filtered, Sterile, Racked (40-384-Tip Racks/Case)
SDP
Supplier:  AUTOMATION TRAINER TM50CLFSR384
Encompass_Preferred
Automation Trainer's Tecan MCA-Compatible 384-Format Pipette Tips are designed to be a direct replacement to Tecan's 384-Format 15 L, 50 L, & 125 L MCA tip. Our tips are manufactured to strict standards to ensure quality and consistent performance. Automation control software protocol modification and separate labware definition is not required.
Catalog No. NC2044889