Learn More
ILEX LIFE SCIENCES LLC 50X HT SUPPLEMENT 6 X 100 ML
SDP
Supplier:  ILEX LIFE SCIENCES LLC P0701100X6
Encompass_Preferred
PAN-Biotech HT Supplement (50x), 6 x 100 ml, Cell Culture Media Supplement, Hypoxanthine + Thymidine. DESCRIPTION: HT Supplement (50x) is a liquid mixture of hypoxanthine and thymidine used as a media supplement in cell culture applications. CONCENTRATION: 50x concentrated. STORAGE: -20°C. STERILE: Yes. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. COMPOSITION: Hypoxanthine: 680.50 mg/L; Thymidine: 388.00 mg/L.
Catalog No. 50-273-3721