Learn More
Sino Biological Recombinant Mouse ICA Protein (His Tag)
Supplier:  Sino Biological 51393M08H100
Encompass_Preferred
A DNA sequence encoding the mouse ICA (NP_082194.1) (Met1-Tyr700) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3100