Learn More
Sino Biological Recombinant Mouse RbAp48 Protein (His Tag)
Supplier:  Sino Biological 51526M07B50
Encompass_Preferred
A DNA sequence encoding the mouse RBBP4 (NP_033056.2) (Met1-Ser425) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-3109