Learn More
eMolecules​ 53292-89-0 | trans-4-(Boc-amino)cyclohexanecarboxylic acid | Oakwood Chemical | MFCD01862293 | 243.303 | C12H21NO4 | 98.000 | CC(C)(C)OC(=O)N[C@H]1CC[C@@H](CC1)C(O)=O | 1g | 537694492
Supplier:  eMolecules​ 537694492
Encompass_Preferred
trans-4-(Boc-amino)cyclohexanecarboxylic acid | Oakwood Chemical | 53292-89-0 | MFCD01862293 | 243.303 | C12H21NO4 | 98.000 | CC(C)(C)OC(=O)N[C@H]1CC[C@@H](CC1)C(O)=O | 1g | 537694492
Catalog No. 50-001-58373