Learn More
ORS Nasco 5400 FULL FACEPIECE SMALL
Supplier:  ORS Nasco 06854001S
Encompass_Preferred
Honeywell part# 54001S. Honeywell North 5400 Series Full Facepiece Air-Purifying Respirator Size: Small ANSI Standard: ANSI Z87.1 Certifications: NIOSH 42 CFR Part 84 OSHA 29 CFR Part 1920.134 Black Full Facepiece Respirator Connection Type: Threaded Model: 54001S
Catalog No. 50-255-1673