Learn More
eMolecules​ 1353892-53-1 | 5,5,5-Trifluoropenylzinc iodide, 0.50 M in THF | Synthonix317.400 | C5H8F3IZn | 0.000 | [I-].FC(F)(F)CCCC[Zn+] | 100ml | 532653003
Supplier:  eMolecules​ 532653003
Encompass_Preferred
5,5,5-Trifluoropenylzinc iodide, 0.50 M in THF | Synthonix | 1353892-53-1317.400 | C5H8F3IZn | 0.000 | [I-].FC(F)(F)CCCC[Zn+] | 100ml | 532653003
Catalog No. 50-269-5995