Learn More
Aat Bioquest 5(6)-FAM [5-(and-6)-Carboxyfluorescein] [CAS 72088-94-9]
SDP
Supplier:  Aat Bioquest 102
Encompass_Preferred
Carboxyfluorescein (commonly called FAM) and its amine-reactive succinimidyl esters are favored over FITC in bioconjugations.
Catalog No. 50-297-9164