Learn More
Aat Bioquest 5(6)-FAM, SE [5-(and-6)-Carboxyfluorescein, succinimidyl ester] [CAS 117548-22-8]
SDP
Supplier:  Aat Bioquest 111
Encompass_Preferred
Carboxyfluorescein (commonly called FAM) and its amine-reactive succinimidyl esters are favored over FITC in bioconjugations.
Catalog No. 50-297-9221