Learn More
Abcam 5,7-Dichlorokynurenic acid sodium salt (mM/ml), NMDA receptor glycine site antagonist, 10MM_ML
Supplier:  Abcam AB14669110MM_ML
Encompass_Preferred
MW 280.04 Da, Purity >99%. Potent NMDA receptor glycine site antagonist. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4525