Learn More
eMolecules​ 59247-47-1 | tert-Butyl 4-bromobenzoate | Combi-Blocks, Inc. | MFCD00093266 | 257.127 | C11H13BrO2 | 98.000 | CC(C)(C)OC(=O)c1ccc(Br)cc1 | 500g | 837443723
Supplier:  eMolecules​ 837443723
Encompass_Preferred
tert-Butyl 4-bromobenzoate | Combi-Blocks, Inc. | 59247-47-1 | MFCD00093266 | 257.127 | C11H13BrO2 | 98.000 | CC(C)(C)OC(=O)c1ccc(Br)cc1 | 500g | 837443723
Catalog No. 50-001-59485