Learn More
ABclonal Technology 594 anti-h CD235a
SDP
Supplier:  ABclonal Technology A25111500T
Encompass_Preferred
Glycophorins A (GYPA) and B (GYPB) are major sialoglycoproteins of the human erythrocyte membrane which bear the antigenic determinants for the MN and Ss blood groups In addition to the M or N and S or s antigens that commonly occur in all populations about 40 related variant phenotypes have been identified These variants include all the variants of the Miltenberger complex and several isoforms of Sta as well as Dantu Sat He Mg and deletion variants Ena S-s-U- and Mk Most of the variants are the result of gene recombinations between GYPA and GYPB
Catalog No. 50-000-08847