Learn More
ABclonal Technology 594 anti-h CD62L/L-Selectin
SDP
Supplier:  ABclonal Technology A24632500T
Encompass_Preferred
This gene encodes a cell surface adhesion molecule that belongs to a family of adhesion homing receptors The encoded protein contains a C-type lectin-like domain a calcium-binding epidermal growth factor-like domain and two short complement-like repeats The gene product is required for binding and subsequent rolling of leucocytes on endothelial cells facilitating their migration into secondary lymphoid organs and inflammation sites Single-nucleotide polymorphisms in this gene have been associated with various diseases including immunoglobulin A nephropathy Alternatively spliced transcript variants have been found for this gene
Catalog No. 50-000-13044