Learn More
ABclonal Technology 594 Rabbit anti-Human CD131
SDP
Supplier:  ABclonal Technology A25076200T
Encompass_Preferred
The protein encoded by this gene is the common beta chain of the high affinity receptor for IL-3 IL-5 and CSF Defects in this gene have been reported to be associated with protein alveolar proteinosis (PAP)
Catalog No. 50-000-12934