Learn More
ABclonal Technology 594 Rabbit anti-Human CD300c
SDP
Supplier:  ABclonal Technology A24099100T
Encompass_Preferred
The CMRF35 antigen which was identified by reactivity with a monoclonal antibody is present on monocytes neutrophils and some T and B lymphocytes (Jackson et al 1992 [PubMed 1349532])
Catalog No. 50-000-07101