Learn More
ABclonal Technology 594 Rabbit anti-Human CD34
SDP
Supplier:  ABclonal Technology A25895200T
Encompass_Preferred
Enables sulfate binding activity Acts upstream of or within leukocyte migration Located in cytoplasm external side of plasma membrane and extracellular region Is expressed in several structures including alimentary system cardiovascular system extraembryonic component genitourinary system and skin Orthologous to human CD34 (CD34 molecule)
Catalog No. 50-000-12631