Learn More
ABclonal Technology 594 Rabbit anti-Human CD5
SDP
Supplier:  ABclonal Technology A24793200T
Encompass_Preferred
This gene encodes a member of the scavenger receptor cysteine-rich (SRCR) superfamily Members of this family are secreted or membrane-anchored proteins mainly found in cells associated with the immune system This protein is a type-I transmembrane glycoprotein found on the surface of thymocytes T lymphocytes and a subset of B lymphocytes The encoded protein contains three SRCR domains and may act as a receptor to regulate T-cell proliferation Alternative splicing results in multiple transcript variants encoding different isoforms
Catalog No. 50-000-11248