Learn More
ABclonal Technology 594 Rabbit anti-Human CD61
SDP
Supplier:  ABclonal Technology A25339500T
Encompass_Preferred
The ITGB3 protein product is the integrin beta chain beta 3 Integrins are integral cell-surface proteins composed of an alpha chain and a beta chain A given chain may combine with multiple partners resulting in different integrins Integrin beta 3 is found along with the alpha IIb chain in platelets Integrins are known to participate in cell adhesion as well as cell-surface mediated signalling
Catalog No. 50-000-10667