Learn More
ABclonal Technology 594 Rabbit anti-Human CD86
SDP
Supplier:  ABclonal Technology A24256100T
Encompass_Preferred
This gene encodes a type I membrane protein that is a member of the immunoglobulin superfamily This protein is expressed by antigen-presenting cells and it is the ligand for two proteins at the cell surface of T cells CD28 antigen and cytotoxic T-lymphocyte-associated protein 4 Binding of this protein with CD28 antigen is a costimulatory signal for activation of the T-cell Binding of this protein with cytotoxic T-lymphocyte-associated protein 4 negatively regulates T-cell activation and diminishes the immune response Alternative splicing results in several transcript variants encoding different isoforms
Catalog No. 50-000-13182