Learn More
ABclonal Technology 594 Rabbit anti-Human CDCP1
SDP
Supplier:  ABclonal Technology A24185200T
Encompass_Preferred
This gene encodes a transmembrane protein which contains three extracellular CUB domains and acts as a substrate for Src family kinases The protein plays a role in the tyrosine phosphorylation-dependent regulation of cellular events that are involved in tumor invasion and metastasis Alternative splicing results in multiple transcript variants of this gene
Catalog No. 50-000-07557