Learn More
ABclonal Technology 594 Rabbit anti-Human IL-17RB
SDP
Supplier:  ABclonal Technology A24317500T
Encompass_Preferred
The protein encoded by this gene is a cytokine receptor This receptor specifically binds to IL17B and IL17E but does not bind to IL17 and IL17C This receptor has been shown to mediate the activation of NF-kappaB and the production of IL8 induced by IL17E The expression of the rat counterpart of this gene was found to be significantly up-regulated during intestinal inflammation which suggested the immunoregulatory activity of this receptor
Catalog No. 50-000-09188