Learn More
ABclonal Technology 594 Rabbit anti-Human IL8
SDP
Supplier:  ABclonal Technology A24961100T
Encompass_Preferred
The protein encoded by this gene is interleukin-8 (IL-8) a major mediator of inflammation and a member of the CXC chemokine family IL-8 is secreted by macrophages neutrophils eosinophils T lymphocytes epithelial cells and fibroblasts guiding neutrophils to infection sites It is rapidly induced by bacterial and viral products and contributes to the proinflammatory cascade playing a role in systemic inflammatory response syndrome (SIRS) IL-8 is involved in conditions like bronchiolitis cystic fibrosis coronary artery disease and endothelial dysfunction It also promotes tumor migration invasion angiogenesis and metastasis IL-8 binds to its receptor (CXCR2) increasing blood vessel permeability Elevated IL-8 levels correlate with worsened disease outcomes including sepsis This gene is part of a CXC chemokine cluster on chromosome 4q
Catalog No. 50-000-12259