Learn More
ABclonal Technology 594 Rabbit anti-Human NG2
SDP
Supplier:  ABclonal Technology A24980500T
Encompass_Preferred
A human melanoma-associated chondroitin sulfate proteoglycan plays a role in stabilizing cell-substratum interactions during early events of melanoma cell spreading on endothelial basement membranes CSPG4 represents an integral membrane chondroitin sulfate proteoglycan expressed by human malignant melanoma cells
Catalog No. 50-000-11975