Learn More
Roboz Surgical #5A OBLIQUE POINT MICRO TWEEZE
SDP
Supplier:  Roboz Surgical RS4928
Encompass
Dumont Tweezers; Pattern #5A; Oblique Points; .06 X .02mm Tip Size; Inox; 115mm Length; Material; Biologie Tips
Catalog No. NC9981249