Learn More
Cayman Chemical 5a-Pregn-3a 20a-dIol 1mg
SDP
Supplier:  Cayman Chemical 388901 MG
Encompass_Preferred
An active metabolite of progesterone; increases intracellular calcium levels in washed isolated human platelets at 10 µM
Catalog No. 50-291-8808