Learn More
Cayman Chemical 5bromo3phenyl SalIcylIc25mg
SDP
Supplier:  Cayman Chemical 1337925 MG
Encompass_Preferred
Selective inhibitor of aldo-keto reductase 1C1 (AKR1C1; Ki = 140 nM) versus AKR1C2 and AKR1C3 (Ki = 1.97 and 21 µM, respectively); does not inhibit AKR1C4 at 100 µM
Catalog No. 50-295-4745