Learn More
Cayman Chemical 5Cyno2 3dI ptolyltetra 10mg
SDP
Supplier:  Cayman Chemical 1592610 MG
Encompass_Preferred
A redox-sensitive tetrazolium salt used primarily to detect metabolic activity in microorganisms; readily taken up by living cells and reduced by respiratory activity to produce a red fluorescent, water-insoluble formazan crystal (absorption maximum at 450 nm)
Catalog No. 50-296-9579