Learn More
CPC Scientific 5FAM-Ala-Glu-Glu-Glu-Tyr-Phe-Phe-Leu-Phe-NH2 (acetate salt) 0.5MG
SDP
Supplier:  CPC Scientific PROK033A
Encompass
ONE-LETTER SEQUENCE: 5FAM-AEEEYFFLF-NH2MOLECULAR FORMULA: C81H86N10O22MOLECULAR WEIGHT:1551.6STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0279