Learn More
CPC Scientific 5FAM-Ala-Lys-Arg-Arg-Arg-Leu-Ser(PO3H2)-Ser(PO3H2)-Leu-Arg-Ala-OH 0.5MG
SDP
Supplier:  CPC Scientific PROK035A
Encompass
SEQUENCE: 5FAM-Ala-Lys-Arg-Arg-Arg-Leu-Ser(PO3H2)-Ser(PO3H2)-Leu-Arg-Ala-OHONE-LETTER SEQUENCE: 5FAM-AKRRRL-Ser(PO3H2)-Ser(PO3H2)-LRAMOLECULAR FORMULA: C75H116N24O26P2MOLECULAR WEIGHT: 1831.9STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES:
SKU(s): PROK-035A, PROK-035B
Catalog No. 50-211-0283