Learn More
Aat Bioquest 5-FITC [Fluorescein-5-isothiocyanate] [CAS 3326-32-7]
SDP
Supplier:  Aat Bioquest 120
Encompass_Preferred
5-FITC and 6-FITC have very similar absorption and fluorescence spectra.
Catalog No. 50-297-9222