Learn More
Cayman Chemical 5fluoro PB22 8hdroxIsoqu 10mg
SDP
Supplier:  Cayman Chemical 1455310 MG
Encompass_Preferred
Differs from 5-fluoro PB-22 by having the quinoline group replaced with an isoquinoline group attached at its eight position; intended for forensic and research purposes
Catalog No. 50-297-1079