Learn More
Medchemexpress LLC 5H-dibenzo[a,d]cyclohepten-10-ol, 10,11-dihydro-5-[3-(methylamino)propylidene]-d3, (5Z)- | 47132-19-4 | 99.0% | 282.40 g/mol | C19H18D3NO | 100 UG

Supplier: Medchemexpress LLC HY100646AS100UG
(Z)-10-Hydroxynortriptyline-d3 is the deuterium-labeled isotopic analogue of the nortriptyline metabolite (Z)-10-hydroxynortriptyline, provided as a high-purity analytical standard for metabolite identification, quantitative LC-MS/MS assays, and pharmacokinetic studies.
- Deuterium-labeled internal standard suitable for LC-MS/MS quantification.
- High purity supporting accurate quantitative analysis and low background.
- Supplied in microgram-scale packaging for trace-level analytical workflows.
- Chemical formula C19H18D3NO and molecular weight 282.40 g/mol aid method setup.
- Stability and handling information provided in accompanying COA and SDS.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.