Learn More
Enzo Life Sciences BIOARRAY™ 5-hmC methylation kit (50tests)
Supplier: Enzo Life Sciences ENZ450110050
The BIOARRAY™ 5-hmC methylation kit allows for sequence specific detection of 5-hydroxymethylcytosine within DNA using a simple and efficient glucosylation reaction procedure.