Learn More
Cayman Chemical 5Hydroxymethyl2furncarbox
SDP
Supplier:  Cayman Chemical 22999500 MG
Encompass_Preferred
A major metabolite of 5-hydroxymethyl-2-furfural; can be used as a building block in the enzymatic synthesis of macrocyclic oligoesters
Catalog No. 50-299-4487