Learn More
Sklar Instruments Single Use, Econo-Sterile, Dressing Forceps, Sklar 5 in., Blunt, Straight, Serrated, Stainless Steel, Disposable, CS of 100.
SDP
Supplier:  Sklar Instruments 962660
Encompass_Preferred
Single Use, Econo-Sterile, Dressing Forceps, Sklar, 5 in., Blunt, Straight, Serrated, Stainless Steel, Disposable, CS of 100. Econo Sterile Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds.
Catalog No. 50-109-4337