Learn More
Cytiva T-series Centrasette cassette Omega PES membrane 5 kDa MWCO 0 5 m
Supplier:  Cytiva OS005T06
Encompass
T-series Centrasette cassette Omega PES membrane 5 kDa MWCO 0 5 m
Catalog No. 50-000-52905