Learn More
NEST SCIENTIFIC USA 5L High Efficiency Erlenmeyer Flask with Bi-directional Transfer Cap, TPE Tube (50cm 1/8" ID 1/4" OD), Vent Filter (0.22 μm), Sterile, 1/pk, 4/cs - C10422-AZB050A
Supplier:  NEST SCIENTIFIC USA C10422AZB050A
Encompass_Preferred
Nest 5L high efficiency shaker flasks are ideal for medium size scale up work. They are "high efficiency" in that a 5L flask has 230mm diameter footprint, allowing for more of them to fit on a shaker platform. The geometry of the flasks also allows for higher shaker speeds without damaging the cells. Faster shaker speeds means better gas exchange, higher growth rates and more yield per flask. This product features pre-assembled tubing, connectors and gas exchange filters allowing for the movement of liquid in and out of the Erlenmeyer flask with minimal risk of contamination. Many customers have realized significant cost savings by switching to aseptic transfer systems. One Nest's largest clients saved over $1m a year by reducing their losses due to cell culture infections.
Catalog No. NC2887695