Learn More
Julabo USA Inc 5m M30 x 1.5 vacuum tubing
SDP
Supplier:  Julabo USA Inc 8891814
Encompass_Preferred
JULABO vacuum-insulated metal tubing is the ideal accessory for high and low-temperature applications including those powered by our PRESTO systems and Ultralow Chillers Temperature range of -100C to 400C Tubing from 0 5m to 5m lengths with M-threaded female fittings The insulated tubing prevents frost build-up and maintains optimal cooling capacity This tubing also increases energy efficiency decreases heat loss and provides better temperature accuracy for faster processing The flexible tubing makes connecting the application to the temperature control unit easy and hassle-free Cleanroom-compatible materials (316L SS) with low spring back that reduces stress on connections and joints Maintains a safer external surface temperature minimizing heat loss Includes one year warranty from ship date
Catalog No. 50-000-77874