Learn More
Cayman Chemical 5methyl7MethoxyIsoflavon 25g
SDP
Supplier:  Cayman Chemical 3926725 G
Encompass_Preferred
A synthetic isoflavone; inhibits the clonogenic survival of HCT116 cells at 10, 20, and 40 µM
Catalog No. 50-291-6579